missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSK2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23762-1-AP-150UL
This item is not returnable.
View return policy
Description
RSK3 is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in the gene have been associated with Coffin-Lowry syndrome (CLS).Specifications
| RSK2 | |
| Polyclonal | |
| Unconjugated | |
| RPS6KA3 | |
| CLS, HU 3, ISPK 1, ISPK1, MAPKAP kinase 1b, MAPKAPK 1b, MAPKAPK1B, MRX19, p90 RSK 3, p90 RSK2, p90RSK3, pp90RSK2, Ribosomal S6 kinase 2, RPS6KA3, RSK, RSK 2, RSK2, S6K alpha 3, S6K alpha3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110651, 6197 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18654, P51812 | |
| RPS6KA3 | |
| RSK2 Fusion Protein Ag20620 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction