missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529280
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, 293T, A431, PC-12, BCL-1. Predicted reactivity: Mouse (100%), Rat (99%), Xenopus laevis (95%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RPS6KA3 (RSK2) is a serine/threonine kinase that modulates many cellular processes including transcription, adhesion, growth, survival, and proliferation. RPS6KA3 contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in the gene have been associated with Coffin-Lowry syndrome (CLS).
Specifications
| RSK2 | |
| Polyclonal | |
| Unconjugated | |
| RPS6KA3 | |
| 90 kDa ribosomal protein S6 kinase 3; C130006E23; CLS; HU-3; Insulin-stimulated protein kinase 1; ISPK1; ISPK-1; MAP kinase-activated protein kinase 1b; MAPK-activated protein kinase 1b; MAPKAP kinase 1b; MAPKAPK1B; MAPKAPK-1b; MPK-9; MRX19; p90-RSK 3; p90-RSK2; p90RSK3; pp90RSK2; RGD1563860; ribosomal protein S6 kinase 2; ribosomal protein S6 kinase A3; ribosomal protein S6 kinase alpha-3; ribosomal protein S6 kinase polypeptide 3; ribosomal protein S6 kinase, 90kDa, polypeptide 3; ribosomal S6 kinase 2; Rps6ka3; Rps6ka-rs1; RSK; Rsk2; RSK-2; S6K-alpha3; S6K-alpha-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110651, 501560, 6197 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P18654, P51812 | |
| RPS6KA3 | |
| Recombinant protein encompassing a sequence within the center region of human RSK2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction