missing translation for 'onlineSavingsMsg'
Learn More

RSK2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18404494 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404494 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404494 Supplier Novus Biologicals Supplier No. NBP130107AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RSK2 Polyclonal antibody specifically detects RSK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSK2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias 90 kDa ribosomal protein S6 kinase 3, CLS, Coffin-Lowry syndrome, EC 2.7.11, EC 2.7.11.1, HU-3, Insulin-stimulated protein kinase 1, ISPK1, ISPK-1, MAP kinase-activated protein kinase 1b, MAPK-activated protein kinase 1b, MAPKAP kinase 1b, MAPKAPK-1b, MAPKAPK1B, mental retardation, X-linked 19, MRX19, p90-RSK2, p90RSK3, pp90RSK2, ribosomal protein S6 kinase alpha-3, ribosomal protein S6 kinase, 90kD, polypeptide 3, ribosomal protein S6 kinase, 90kDa, polypeptide 3, Ribosomal S6 kinase 2, RSK, RSK-2, RSK2p90-RSK 3, S6K-alpha3, S6K-alpha-3
Host Species Rabbit
Immunogen maps to a region between residue 1 and 50 of human ribosomal S6 kinase 2 using the numbering given in entry NP_004577.1
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6197
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.