missing translation for 'onlineSavingsMsg'
Learn More

RSK1, p Ser380 Antibody, Novus Biologicals™

Product Code. 18370826 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370826 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370826 Supplier Novus Biologicals Supplier No. NBP313243

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RSK1 Polyclonal antibody specifically detects RSK1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 1% BSA, 20% Glycerol (pH7)
Gene Alias 90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide corresponding to residues around human RSK1 (phospho Ser380). The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6195
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.