missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Rsk1/MAPKAP Kinase 1a Protein, Active

Product Code. 17360224
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17360224

Brand: Eurofins DiscoverX 14479

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, recombinant full-length rat RSK1 (residues 1-end), containing additional N-terminal His6 and HA (YPYDV PDYA) tags, for use in Kinase Assays.

Rsk1, also known as Ribosomal Protein S6 Kinase A1, is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

GenBank M99169

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 89.9 kDa
Quantity 10 μg
Species Rat
Recombinant Recombinant
Protein Tag N-terminal 6His and N-terminal HA (YPYDVPDYA)
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥72%
Protein Rsk1/MAPKAP Kinase 1a, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.