missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSG1 Antibody (3G4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079363-M02
This item is not returnable.
View return policy
Description
RSG1 Monoclonal antibody specifically detects RSG1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RSG1 | |
| Monoclonal | |
| Unconjugated | |
| AAH02946 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3G4 | |
| In 1x PBS, pH 7.4 | |
| C1orf89, chromosome 1 open reading frame 89, MGC10731, miro domain-containing protein C1orf89, Rem/Rab-Similar GTPase 1, REM2 and RAB-like small GTPase 1, RP4-733M16.4 | |
| C1orf89 (AAH02946.1, 1 a.a. ∽ 258 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MARPPVPGSVVVPNWHESAEGKEYLACILRKNRRRVFGLLERPVLLPPVSIDTASYKIFVSGKSGVGKTALVAKLAGLEVPVVHHGTTGIQTTVVFWPAKLQASSRVVMFRFEFWDCGESALKKFDHMLLACMENTDAFLFLFSFTDRASFEDLPGQLARIAGEAPGVVRMVIGSKFDQYMHTDVPERDLTAFRQAWELPLLRVKSVPGRRLADGRTLDGRAGLADVAHILNGLAEQLWHQDQVAAGLLPNPPESAPE | |
| 0.1 mg | |
| Signal Transduction | |
| 79363 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction