missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583806
This item is not returnable.
View return policy
Description
Immunogen sequence: KYLCECQFDD NLKFKNIINE EDADTMRLQP IGRDKDGLMY WYQLDQDHNV RMYIEEQDDQ DGSSWKCIVR NRNELAETLA LLKAQIDPVL LK Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 100%.
RSF1 is required for assembly of regular nucleosome arrays by the RSF chromatin remodelling complex. Rsf1 facilitates transcription of hepatitis B virus (HBV) genes by the pX transcription activator. In case of infection by HBV, together with pX, it represses TNF-alpha induced NF-kappaB transcription activation. Rsf1 represses transcription when artificially recruited to chromatin by fusion to a heterogeneous DNA binding domain. Rsf1 interacts with SMARCA5/SNF2H to form the RSF complex and also binds the HBV pX/HBx protein, which is required to activate transcription of the viral genome.
Specifications
| RSF1 | |
| Polyclonal | |
| Unconjugated | |
| RSF1 | |
| 4832420A03Rik; C030033M12Rik; Gm164; HBV pX associated protein-8; HBV pX-associated protein 8; HBXAP; hepatitis B virus x associated protein; hepatitis B virus x-associated protein; p325; p325 subunit of RSF chromatin-remodeling complex; Remodeling and spacing factor 1; RSF1; RSF-1; XAP8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51773 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96T23 | |
| RSF1 | |
| Recombinant protein corresponding to Human RSF1. Recombinant protein control fragment (Product #RP-106109). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction