missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSAD2 Mouse, anti-Human, Clone: 1C5D4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RSAD2 |
|---|---|
| Clone | 1C5D4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18020626
|
Proteintech
68604-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18085285
|
Proteintech
68604-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RSAD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RSAD2. This antibody recognizes human, mouse, pig antigen. The RSAD2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RSAD2 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Pig | |
| 2510004L01Rik, cig33, cig5, RSAD2, vig1, Viperin | |
| Primary |
| 1C5D4 | |
| Western Blot, ELISA | |
| Mouse | |
| 91543 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title