missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSAD1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA525186

Product Code. 13262989

  • £440.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

RSAD1 may be involved in porphyrin cofactor biosynthesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RSAD1
Polyclonal
Unconjugated
RSAD1
B430319G23; BC056485; oxygen-independent coproporphyrinogen-III oxidase-like protein RSAD1; Putative heme chaperone; radical S-adenosyl methionine domain containing 1; radical S-adenosyl methionine domain-containing protein 1, mitochondrial; RSAD1
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
55316
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9HA92
RSAD1
KLH conjugated synthetic peptide between 299-328 amino acids from the C-terminal region of human RSAD1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.