missing translation for 'onlineSavingsMsg'
Learn More

RS102895 hydrochloride, MedChemExpress

Product Code. 16453557
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453557 50 mg 50mg
16437538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16453557 Supplier Medchem Express Supplier No. HY18611
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress RS102895 hydrochloride is a potent CCR2 antagonist, with an IC50 of 360 nM, and shows no effect on CCR1.

RS102895 hydrochloride is a potent CCR2 antagonist, with an IC50 of 360 nM, and shows no effect on CCR1.

Specifications

Chemical Name or Material RS102895 hydrochloride
CAS 1173022-16-6
Color White
Purity Grade Notes Research
Molecular Formula C21H22ClF3N2O2
Quantity 50 mg
Solubility Information DMSO : 41.67 mg/mL (97.62 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [H]Cl.O=C1NC2=CC=CC=C2C3(CCN(CCC4=CC=C(C(F)(F)F)C=C4)CC3)O1
Molecular Weight (g/mol) 426.86
Formula Weight 426.86
Percent Purity 98.32%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.