missing translation for 'onlineSavingsMsg'
Learn More

(RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-(3-fluorophenylsulfonyl)-2-hydroxy-2-methylpropanamide, Mikromol™

Product Code. 30049707
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30049707 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 30049707

Brand: Mikromol MM0894.080025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-(3-fluorophenylsulfonyl)-2-hydroxy-2-methylpropanamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-(3-fluorophenylsulfonyl)-2-hydroxy-2-methylpropanamide
CAS 1166228-30-3
Analyte or Component Names (RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-(3-fluorophenylsulfonyl)-2-hydroxy-2-methylpropanamide
Shipping Condition Room Temperature
Molecular Formula C18 H14 F4 N2 O4 S
Certifications/Compliance ISO 17025
Synonym 3-Fluorophenyl Bicalutamide, (RS)-N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(3-fluorophenylsulfonyl)-2-hydroxy-2-methylpropanamide, 3-Fluorophenyl Bicalutamide, Bicalutamide Related Compound B (USP), Bicalutamide Related Compound B
InChI Formula InChI=1S/C18H14F4N2O4S/c1-17(26,10-29(27,28)14-4-2-3-12(19)7-14)16(25)24-13-6-5-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)
SMILES CC(O)(CS(=O)(=O)c1cccc(F)c1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(3-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
Molecular Weight (g/mol) 430.37
Quantity 25 mg
Formula Weight 430.061 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.