missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (RS)-MCPG disodium salt

Description
(RS)-MCPG disodium salt is a sodium salt of (RS)-MCPG, a non-selective group I/group II metabotropic glutamate receptor antagonist.
S-enantiomer also available.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | (RS)-MCPG disodium salt |
| Purity | >99% |
| Molecular Weight (g/mol) | 253.16 |
| Inhibitors | (RS)-MCPG disodium salt |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?