missing translation for 'onlineSavingsMsg'
Learn More

(RS)-Equol, Mikromol™

Product Code. 30051810
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051810 25 mg 25mg
17949194 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051810 Supplier Mikromol Supplier No. MM0747.020025
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(RS)-Equol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material R,S-Equol
CAS 94105-90-5
Analyte or Component Names R,S-Equol
Shipping Condition Room Temperature
Molecular Formula C15 H14 O3
Certifications/Compliance ISO 17025
Synonym 2H-1-Benzopyran-7-ol, 3,4-dihydro-3-(4-hydroxyphenyl)-, 7,4'-Homoisoflavane, (+/-)-Equol, NV 07alpha
InChI Formula InChI=1S/C15H14O3/c16-13-4-1-10(2-5-13)12-7-11-3-6-14(17)8-15(11)18-9-12/h1-6,8,12,16-17H,7,9H2
SMILES Oc1ccc(cc1)C2COc3cc(O)ccc3C2
IUPAC Name 3-(4-hydroxyphenyl)-3,4-dihydro-2H-chromen-7-ol
Molecular Weight (g/mol) 242.27
Quantity 25 mg
Formula Weight 242.0943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.