Learn More
BINDER™ RS 485 / RS 422 Interface Converter
Product Code. 15974252 Shop All BINDER Products
Click to view available options
Material:
Silicone
Unit Size:
Each
Product Code. 15974252
Brand: BINDER™ 80120589
Only null left
Add to basket
Add to basket
For Use With: All MKF, MKT, MKFT Series models
  • RS 422 cable set and RS 485 / RS 422 interface converter for connection to 10-way plug distributor
Specifications
Description RS 485 / RS 422 interface converter - 230 V option model, RS 422 cable set and RS 485 / RS 422 interface converter for connection to 10-way plug distributor
For Use With (Equipment) BINDER Chambers Series MKF, MKT, MKFT
Material Silicone
Type Interface Converter