missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RS 23597-190 hydrochloride
GREENER_CHOICE

Product Code. 18005162
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18005162 50 mg 50mg
30001117 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18005162 Supplier Tocris Bioscience™ Supplier No. 0728/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-HT4 antagonist

A high affinity, selective competitive antagonist at 5-HT4 receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149719-06-2
Molecular Formula C16H24Cl2N2O3

Specifications

Quantity 50 mg
Formula Weight 363.28
Purity >98%
Molecular Weight (g/mol) 363.279
Chemical Name or Material RS 23597-190 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.