missing translation for 'onlineSavingsMsg'
Learn More

(RS)-20,22-Dihydrodigoxigenin, TRC

Product Code. 30379096
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379096 25 mg 25mg
30370873 2.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379096 Supplier Toronto Research Chemicals Supplier No. TRCD45229025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105880-19-1
Molecular Formula C23H36O5
Molecular Weight (g/mol) 392.53
Synonym Dihydrodigoxigenin, 20,22-Dihydro-digoxigenin, 3β,12β,14-Trihydroxy-5β-cardanolide, (3β,5β,12β)-3,12,14-trihydroxy-cardanolide
IUPAC Name 4-((3S,5R,8R,9S,10S,12R,13S,14S,17R)-3,12,14-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)dihydrofuran-2(3H)-one
SMILES O[C@]([C@@](CC[C@@]1([H])[C@@]2(CC[C@H](O)C1)C)([H])[C@]2([H])C[C@H]3O)(CC[C@]4([H])C(C5)([H])COC5=O)[C@]34C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H36O5/c1-21-7-5-15(24)10-14(21)3-4-17-18(21)11-19(25)22(2)16(6-8-23(17,22)27)13-9-20(26)28-12-13/h13-19,24-25,27H,3-12H2,1-2H3/t13?,14-,15+,16-,17-,18+,19-,21?,22?,23+/m1/s1
Formula Weight 392.26
Chemical Name or Material (RS)-20,22-Dihydrodigoxigenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.