missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(RS)-2-Chloro-9-[3-(dimethylamino)propyl]-9H-thioxanthen-9-ol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(RS)-2-Chloro-9-[3-(dimethylamino)propyl]-9H-thioxanthen-9-ol
Specifications
Specifications
| Chemical Name or Material | (RS)-2-Chloro-9-[3-(dimethylamino)propyl]-9H-thioxanthen-9-ol |
| CAS | 4295-65-2 |
| Analyte or Component Names | (RS)-2-Chloro-9-[3-(dimethylamino)propyl]-9H-thioxanthen-9-ol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H20 Cl N O S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Chlorprothixene Hydrochloride Imp. A (EP), Chlorprothixene Imp. A (EP), (RS)-2-Chloro-9-[3-(dimethylamino)propyl]-9H-thioxanthen-9-ol, Chlorprothixene Hydrochloride Impurity A, Chlorprothixene Impurity A |
| InChI Formula | InChI=1S/C18H20ClNOS/c1-20(2)11-5-10-18(21)14-6-3-4-7-16(14)22-17-9-8-13(19)12-15(17)18/h3-4,6-9,12,21H,5,10-11H2,1-2H3 |
| SMILES | CN(C)CCCC1(O)c2ccccc2Sc3ccc(Cl)cc13 |
| IUPAC Name | 2-chloro-9-[3-(dimethylamino)propyl]thioxanthen-9-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction