missing translation for 'onlineSavingsMsg'
Learn More

(RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]-acetic Acid Dihydrochloride (Ethoxycetirizine Dihydrochloride), Mikromol™

Product Code. 17982136
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982136 100 mg 100mg
30051916 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17982136 Supplier Mikromol Supplier No. MM0380.05
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]-acetic Acid Dihydrochloride (Ethoxycetirizine Dihydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]-acetic Acid Dihydrochloride (Ethoxycetirizine Dihydrochloride)
CAS 83881-56-5
Analyte or Component Names (RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]-acetic Acid Dihydrochloride (Ethoxycetirizine Dihydrochloride)
Shipping Condition Room Temperature
Molecular Formula C23 H29 Cl N2 O4 . 2 Cl H
Certifications/Compliance ISO 9001
Synonym Cetirizine Dihydrochloride Imp. E (EP), Cetirizine Imp. E (EP), (RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]-acetic Acid Dihydrochloride, Cetirizine Dihydrochloride Imp. E (EP) as Dihydrochloride, Ethoxycetirizine Dihydrochloride, Cetirizine Dihydrochloride Impurity E as Dihydrochloride, Cetirizine Impurity E as Dihydrochloride
InChI Formula InChI=1S/C23H29ClN2O4.2ClH/c24-21-8-6-20(7-9-21)23(19-4-2-1-3-5-19)26-12-10-25(11-13-26)14-15-29-16-17-30-18-22(27)28;;/h1-9,23H,10-18H2,(H,27,28);2*1H
SMILES Cl.Cl.OC(=O)COCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC Name 2-[2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]acetic acid;dihydrochloride
Molecular Weight (g/mol) 505.86
Quantity 100 mg
Formula Weight 504.1349 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.