missing translation for 'onlineSavingsMsg'
Learn More

RS-127445 hydrochloride, MedChemExpress

Product Code. p-200097281
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16449087

Brand: Medchem Express HY15419

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RS-127445 hydrochloride is a selective, high affinity, orally bioavailable 5-HT2B receptor antagonist with a pKi of 9.5. RS-127445 hydrochloride shows 1000 fold selectivity for this receptor as compared to numerous other receptor and ion channel binding sites.

RS-127445 hydrochloride is a selective, high affinity, orally bioavailable 5-HT2B receptor antagonist with a pKi of 9.5. RS-127445 hydrochloride shows 1000 fold selectivity for this receptor as compared to numerous other receptor and ion channel binding sites.

Specifications

Chemical Name or Material RS-127445 hydrochloride
CAS 199864-86-3
Color White
Purity Grade Notes Research
Molecular Formula C17H17ClFN3
Quantity 5 mg
Solubility Information DMSO : ≥ 31 mg/mL (97.55 mM) ∣H2O : 2.27 mg/mL (7.14 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=NC(C(C)C)=CC(C2=C3C=CC=CC3=C(F)C=C2)=N1.[H]Cl
Molecular Weight (g/mol) 317.79
Formula Weight 317.79
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.