missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RS 102895 hydrochloride
GREENER_CHOICE

Product Code. 18054122
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18054122 10 mg 10mg
18004222 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18054122 Supplier Tocris Bioscience™ Supplier No. 2089/10
  • ONLINE EXCLUSIVE PRICE
    £170.00
    £160.65 / 10mg
    Save £9.35
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

CCR2b chemokine receptor antagonist

CCR2-selective chemokine receptor antagonist (IC50 values are 0.36 and 17.8 μM for inhibition of human recombinant CCR2b and CCR1 receptors respectively). Blocks MCP-1-stimulated calcium influx and chemotaxis with IC50 values of 32 nM and 1.7 μM respectively. Also inhibits α1A, α1D and 5-HT1A receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173022-16-6
Molecular Formula C21H22ClF3N2O2

Specifications

Quantity 10 mg
Formula Weight 426.86
Purity >99%
Molecular Weight (g/mol) 426.864
Chemical Name or Material RS 102895 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.