missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRS1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00023212-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MEGQSVEELLAKAEQDEAEKLQRITVHKELELQFDLGNLLASDRNPPTGLRCAGPTPEAELQALARDNTQLLINQLWQLPTERVEEAIVARLPEPTTRLPREKPLPRPRPLTRWQQFARLKGIRPKKKTNLVWDEVSGQWRRRWGYQRARDDTKEWLIEVPGNADPLEDQFAKRIQAKKERVAKNELNRLRNLARAHKMQLPSAAGLHPTGHQSKEELGRAMQVAKVSTASVGRFQERLPKEKVPRGSGKKRKFQPLFGDFAAEKKNQLELLRVMNSKKPQLDVTRATNKQMREEDQEEAAKRRKMSQKGKRKGGRQGPGGKRKGGPPSQGGKRKGGLGGKMNSGPPGLGGKRKGGQRPGGKRRKSpecifications
| RRS1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_015169 | |
| RRS1 | |
| RRS1 (NP_055984.1, 1 a.a. ∼ 365 a.a) full-length human protein. | |
| RUO | |
| 23212 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| RRS1 ribosome biogenesis regulator homolog (S. cerevisiae) | |
| KIAA0112 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu