missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RRS1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16613165 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16613165 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16613165 Supplier Invitrogen™ Supplier No. PA5106724

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RRS1.

Involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRS1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rrs1
Gene Accession No. A1A5P2, Q15050, Q9CYH6
Gene Alias 5730466A07Rik; AA415037; D1Ertd701e; homolog of yeast ribosome biogenesis regulator; homolog of yeast ribosome biogenesis regulatory protein RRS1; KIAA0112; MNCb-2643; regulator for ribosome resistance homolog; ribosome biogenesis regulator homolog; ribosome biogenesis regulatory protein homolog; ribosome biogenesis regulatory protein RRS1 homolog; Rrr; Rrs1; RRS1 ribosome biogenesis regulator homolog; RRS1 ribosome biogenesis regulator homolog (S. cerevisiae)
Gene Symbols Rrs1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RRS1(Accession Q15050), corresponding to amino acid residues K253-R303.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23212, 297784, 59014
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.