missing translation for 'onlineSavingsMsg'
Learn More

RRS1 Antibody, Novus Biologicals™

Product Code. 18358068 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358068 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358068 Supplier Novus Biologicals Supplier No. H00023212D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RRS1 Polyclonal antibody specifically detects RRS1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRS1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_055984.1
Gene Alias FSDR2, homolog of yeast ribosome biogenesis regulator, homolog of yeast ribosome biogenesis regulatory protein RRS1, KIAA0112ribosome biogenesis regulatory protein RRS1 homolog, ribosome biogenesis regulatory protein homolog, RRR, RRS1 ribosome biogenesis regulator homolog (S. cerevisiae)
Host Species Rabbit
Immunogen RRS1 (NP_055984.1, 1 a.a. - 365 a.a.) full-length human protein. MEGQSVEELLAKAEQDEAEKLQRITVHKELELQFDLGNLLASDRNPPTGLRCAGPTPEAELQALARDNTQLLINQLWQLPTERVEEAIVARLPEPTTRLPREKPLPRPRPLTRWQQFARLKGIRPKKKTNLVWDEVSGQWRRRWGYQRARDDTKEWLIEVPGNADPLEDQFAKRIQAKKERVAKNELNRLRNLARAHKMQLPSAAGLHPTGHQSKEELGRAMQVAKVSTASVGRFQERLPKEKVPRGSGKKRKFQPLFGDFAAEKKNQLELLRVMNSKKPQLDVTRATNKQMREEDQEEAAKRRKMSQKGKRKGGRQGPGGKRKGGPPSQGGKRKGGLGGKMNSGPPGLGGKRKGGQRPGGKRRK
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23212
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.