missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30725-25ul
This item is not returnable.
View return policy
Description
RRS1 Polyclonal specifically detects RRS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RRS1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunofluorescence | |
| Q15050 | |
| RRS1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LHPTGHQSKEELGRAMQVAKVSTASVGRFQERLPKEKVPRGSGKKRKFQPLFGDFAAEKKNQLELLRVMNSKKP | |
| 25 μL | |
| Stem Cell Markers | |
| 23212 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FSDR2, homolog of yeast ribosome biogenesis regulator, homolog of yeast ribosome biogenesis regulatory protein RRS1, KIAA0112ribosome biogenesis regulatory protein RRS1 homolog, ribosome biogenesis regulatory protein homolog, RRR, RRS1 ribosome biogenesis regulator homolog (S. cerevisiae) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction