missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRP7A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100809
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RRP7.
RRP7A (Ribosomal RNA Processing 7 Homolog A) is a Protein Coding gene. Diseases associated with RRP7A include Autosomal Dominant Non-Syndromic Intellectual Disability 1 and Autosomal Dominant Non-Syndromic Intellectual Disability. Among its related pathways are rRNA processing in the nucleus and cytosol and Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleotide binding. [GeneCards].
Specifications
| RRP7A | |
| Polyclonal | |
| Unconjugated | |
| RRP7A | |
| 1110014J01Rik; AA408146; BK126B4.3; CGI-96; CTA-126B4.5; gastric cancer antigen Zg14; Gastric cancer antigen Zg14 homolog; hypothetical protein LOC550241; Kheg1; kidney highly expressed protein 1; RGD1311547; ribosomal RNA processing 7 homolog; ribosomal RNA processing 7 homolog A; ribosomal RNA processing 7 homolog A (S. cerevisiae); ribosomal RNA-processing protein 7 homolog A; RRP7; RRP7A; zgc:110084 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27341, 362967, 74778 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9D1C9, Q9Y3A4 | |
| RRP7A | |
| A synthesized peptide derived from human RRP7A(Accession Q9Y3A4), corresponding to amino acid residues T71-V121. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction