missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RRP7A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA551604

Product Code. 15729560

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VRQGTKSTWP QKRTLFVLNV PPYCTEESLS RLLSTCGLVQ SVELQEKPDL AESPKESRSK FFHPKPVPGF QVAYVVFQKP SGVSAALALK GPLLVSTESH PVKSGIHKWI SDYA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

RRP7A (Ribosomal RNA Processing 7 Homolog A) is a Protein Coding gene. Diseases associated with RRP7A include Autosomal Dominant Non-Syndromic Intellectual Disability 1 and Autosomal Dominant Non-Syndromic Intellectual Disability. Among its related pathways are rRNA processing in the nucleus and cytosol and Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleotide binding. [GeneCards].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RRP7A
Polyclonal
Unconjugated
RRP7A
1110014J01Rik; AA408146; BK126B4.3; CGI-96; CTA-126B4.5; gastric cancer antigen Zg14; Gastric cancer antigen Zg14 homolog; hypothetical protein LOC550241; Kheg1; kidney highly expressed protein 1; RGD1311547; ribosomal RNA processing 7 homolog; ribosomal RNA processing 7 homolog A; ribosomal RNA processing 7 homolog A (S. cerevisiae); ribosomal RNA-processing protein 7 homolog A; RRP7; RRP7A; zgc:110084
Rabbit
Antigen affinity chromatography
RUO
27341
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y3A4
RRP7A
Recombinant protein corresponding to Human RRP7A. Recombinant protein control fragment (Product #RP-91482).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.