missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35142-100ul
This item is not returnable.
View return policy
Description
RRP4 Polyclonal antibody specifically detects RRP4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| RRP4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| exosome component 2Ribosomal RNA-processing protein 4, homolog of yeast RRP4 (ribosomal RNA processing 4), 3' 5' exoribonuclease(RRP4), homolog of yeast RRP4 (ribosomal RNA processing 4), 3'-5'-exoribonuclease, hRrp4p, p7, RRP4exosome complex exonuclease RRP4, Rrp4p | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-293 of human RRP4 (NP_055100.2).,, Sequence:, MAMEMRLPVARKPLSERLGRDTKKHLVVPGDTITTDTGFMRGHGTYMGEEKLIASVAGSVERVNKLICVKALKTRYIGEVGDIVVGRITEVQQKRWKVETNSRLDSVLLLSSMNLPGGELRRRSAEDELAMRGFLQEGDLISAEVQAVFSDGAVSLHTRSLKYGKLGQGVLVQVSPSLVKRQKTHFHDLPCGASVILGNNGFIWIYPTPEHKEEEAGGFIANLEPVSLADREVISRLRNCIISLVTQRMMLYDTSILYCYEASLPHQIKDILKPEIMEEIVMETRQRLLEQEG | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23404 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction