missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP1B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27723-1-AP-150UL
This item is not returnable.
View return policy
Description
RRP1B belongs to the RRP1 family. It may be a novel susceptibility gene for breast cancer progression and metastasis.Specifications
| RRP1B | |
| Polyclonal | |
| Unconjugated | |
| RRP1B | |
| KIAA0179, Nnp1, NNP1L, RRP1, RRP1 like protein B, RRP1B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23076 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14684 | |
| RRP1B | |
| RRP1B Fusion Protein Ag23405 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction