missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530279
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is the putative homolog of the yeast ribosomal RNA processing protein RRP1. The encoded protein is involved in the late stages of nucleologenesis at the end of mitosis, and may be required for the generation of 28S rRNA.
Specifications
| RRP1 | |
| Polyclonal | |
| Unconjugated | |
| RRP1 | |
| AL033305; AL033306; AU018604; D21S2056E; NNP1; NNP-1; Nnp1 homolog, nucleolar protein; NOP52; Novel nuclear protein 1; nucleolar protein Nop52; protein NNP-1; ribosomal RNA processing 1; ribosomal RNA processing 1 homolog; ribosomal RNA processing 1 homolog (S. cerevisiae); ribosomal RNA processing protein 1 homolog A; RRP1; RRP1A; RRP1-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8568 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.64 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P56182 | |
| RRP1 | |
| Recombinant protein encompassing a sequence within the center region of human RRP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction