missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRN3 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71822G
This item is not returnable.
View return policy
Description
RRN3 Polyclonal antibody specifically detects RRN3 in Human samples. It is validated for Western Blot
Specifications
| RRN3 | |
| Polyclonal | |
| Western Blot | |
| DKFZp566E104, RNA polymerase I-specific transcription initiation factor RRN3, RRN3 RNA polymerase I transcription factor homolog (S. cerevisiae), RRN3 RNA polymerase I transcription factor homolog (yeast), TIF-IA, TIFIAMGC104238, Transcription initiation factor IA, transcription initiation factor TIF-IA | |
| The immunogen for this product maps to a region between residue 601 and 651 of human Transcription Initiation Factor IA using the numbering given in entry NP_060897.3 (GeneID 54700). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54700 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction