missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRM2 Recombinant Rabbit Monoclonal Antibody (SAIC-30E-7)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551845
This item is not returnable.
View return policy
Description
RRM2 Recombinant Monoclonal Antibody for MS, ICC/IF, ELISA
RRM2 is one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the protein (M2) is regulated in a cell-cycle dependent fashion.
Specifications
| RRM2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P31350 | |
| RRM2 | |
| The KLH-conjugated peptide DIQHWESLKPEER derived from Ribonucleoside-diphosphate reductase subunit M2 (RRM2). RRM2 provides precursors required for DNA synthesis (aa 112-124). | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Immunocytochemistry, Mass Spectrometry | |
| SAIC-30E-7 | |
| Unconjugated | |
| RRM2 | |
| AA407299; R2; ribonucleoside-diphosphate reductase M2 chain; ribonucleoside-diphosphate reductase subunit M2; Ribonucleotide reductase 2; ribonucleotide reductase M2; ribonucleotide reductase M2 polypeptide; ribonucleotide reductase regulatory subunit M2; ribonucleotide reductase small chain; Ribonucleotide reductase small subunit; RR2; RR2M; RRM2; RRR2M | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6241 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction