missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584324
This item is not returnable.
View return policy
Description
Immunogen sequence: LAPITDPQQL QLSPLKGLSL VDKENTPPAL SGTRVLASKT ARRIFQEPTE PKTKAAAPGV EDEPLLRE Highest antigen sequence indentity to the following orthologs: Mouse - 72%, Rat - 68%.
RRM2 is one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the protein (M2) is regulated in a cell-cycle dependent fashion.
Specifications
| RRM2 | |
| Polyclonal | |
| Unconjugated | |
| RRM2 | |
| AA407299; R2; ribonucleoside-diphosphate reductase M2 chain; ribonucleoside-diphosphate reductase subunit M2; Ribonucleotide reductase 2; ribonucleotide reductase M2; ribonucleotide reductase M2 polypeptide; ribonucleotide reductase regulatory subunit M2; ribonucleotide reductase small chain; Ribonucleotide reductase small subunit; RR2; RR2M; RRM2; RRR2M | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6241 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P31350 | |
| RRM2 | |
| Recombinant protein corresponding to Human RRM2. Recombinant protein control fragment (Product #RP-99902). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction