Learn More
RRM2 Mouse anti-Human, Unconjugated, Clone: 2A9A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67006-1-IG-150UL
This item is not returnable.
View return policy
Description
The RRM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RRM2. This antibody recognizes human antigen. The RRM2 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
| RRM2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunoprecipitation, Immunofluorescence, ELISA | |
| 2A9A7 | |
| Unconjugated | |
| 150 μL | |
| 6241 | |
| IgG1 |