missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRM2 Antibody (SAIC-30E-7) - Chimeric, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43533
This item is not returnable.
View return policy
Description
RRM2 Monoclonal antibody specifically detects RRM2 in Human samples. It is validated for ELISA
Specifications
| RRM2 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA | |
| EC 1.17.4.1, R2, ribonucleoside-diphosphate reductase subunit M2, ribonucleotide reductase M2, ribonucleotide reductase M2 polypeptide, Ribonucleotide reductase small chain, Ribonucleotide reductase small subunit, RR2, RR2M | |
| The KLH-conjugated peptide DIQHWESLKPEER derived from Ribonucleoside-diphosphate reductase subunit M2 (RRM2). RRM2 provides precursors required for DNA synthesis. (Uniprot# P31350) | |
| 0.2 mg | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 6241 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| SAIC-30E-7 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction