missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRM1 Rabbit anti-Human, Clone: RRM1/4372R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | RRM1 |
|---|---|
| Clone | RRM1/4372R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | ELISA, Peptide Array |
Description
RRM1 Monoclonal specifically detects RRM1 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| RRM1 | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 6240 | |
| IgG | |
| Protein A or G purified |
| RRM1/4372R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1 | |
| Synthetic peptide corresponding to residues within aa700-800 of human RRM1 (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title