missing translation for 'onlineSavingsMsg'
Learn More

RRM1 Antibody (2D11), Novus Biologicals™

Product Code. 18356899 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356899 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356899 Supplier Novus Biologicals Supplier No. H00006240M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RRM1 Monoclonal antibody specifically detects RRM1 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRM1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2D11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001024.1
Gene Alias EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1
Host Species Mouse
Immunogen RRM1 (NP_001024.1, 593 a.a. ~ 792 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IRNSLLIAPMPTASTAQILGNNESIEPYTSNIYTRRVLSGEFQIVNPHLLKDLTERGLWHEEMKNQIIACNGSIQSIPEIPDDLKQLYKTVWEISQKTVLKMAAERGAFIDQSQSLNIHIAEPNYGKLTSMHFYGWKQGLKTGMYYLRTRPAANPIQFTLNKEKLKDKEKVSKEEEEKERNTAAMVCSLENRDECLMCGS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6240
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.