missing translation for 'onlineSavingsMsg'
Learn More

RREB1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Product Code. 18040556
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18040556 20 μg 20µL
18384612 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18040556 Supplier Bio-Techne Supplier No. NBP31597220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RREB1 Polyclonal antibody specifically detects RREB1 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RREB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000
Formulation PBS, 50% glycerol, pH7.3
Gene Alias DNA-binding protein, FINB, Finger protein in nuclear bodies, hindsight homolog, HNT, LZ321, Raf-responsive zinc finger protein LZ321, ras responsive element binding protein 1, ras-responsive element-binding protein 1, RREB-1, Zep-1, zinc finger motif enhancer binding protein 1, Zinc finger motif enhancer-binding protein 1, zinc finger motif-enhancer binding-protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-480 of human RREB1 (NP_001003698.1). ATPLPGDALDQKGFLALLGLQHTKDVRPAPAEEPLPDDNQAIQLQTLKCQLPQDPGCTNLLSLSPFEAASLGGSLTVLPATKDSIKHLSLQPFQKGFIIQPDSSIVVKPISGESAIELADIQQILKMAASA
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6239
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.