missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RREB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | RREB1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18245533
|
Novus Biologicals
NBP2-56824 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658917
|
Novus Biologicals
NBP2-56824-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RREB1 Polyclonal specifically detects RREB1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RREB1 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 6239 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GSVPKAATTATPAATTSPKESSEPPAPASSPEAASPTEQGPAGTSKKRGRKRGMRSRPRANSGGVDLDSSGEFASIEKMLATTDTNKFSPF | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| DNA-binding protein, FINB, Finger protein in nuclear bodies, hindsight homolog, HNT, LZ321, Raf-responsive zinc finger protein LZ321, ras responsive element binding protein 1, ras-responsive element-binding protein 1, RREB-1, Zep-1, zinc finger motif enhancer binding protein 1, Zinc finger motif enhancer-binding protein 1, zinc finger motif-enhancer binding-protein 1 | |
| RREB1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title