missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RREB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12657-100ul
This item is not returnable.
View return policy
Description
RREB1 Polyclonal antibody specifically detects RREB1 in Human samples. It is validated for Western Blot
Specifications
| RREB1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DNA-binding protein, FINB, Finger protein in nuclear bodies, hindsight homolog, HNT, LZ321, Raf-responsive zinc finger protein LZ321, ras responsive element binding protein 1, ras-responsive element-binding protein 1, RREB-1, Zep-1, zinc finger motif enhancer binding protein 1, Zinc finger motif enhancer-binding protein 1, zinc finger motif-enhancer binding-protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human RREB1 fragment | |
| 100 μg | |
| Signal Transduction | |
| 6239 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction