missing translation for 'onlineSavingsMsg'
Learn More

RREB1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18407822 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407822 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407822 Supplier Novus Biologicals Supplier No. NBP171884AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RREB1 Polyclonal antibody specifically detects RREB1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RREB1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DNA-binding protein, FINB, Finger protein in nuclear bodies, hindsight homolog, HNT, LZ321, Raf-responsive zinc finger protein LZ321, ras responsive element binding protein 1, ras-responsive element-binding protein 1, RREB-1, Zep-1, zinc finger motif enhancer binding protein 1, Zinc finger motif enhancer-binding protein 1, zinc finger motif-enhancer binding-protein 1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1375 and 1425 of human Ras Responsive Eement Binding Protein 1 using the numbering given in entry NP_002946.4 (GeneID 6239).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6239
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.