missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RREB1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-71884AF532
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
RREB1 Polyclonal antibody specifically detects RREB1 in Human samples. It is validated for Western Blot
Especificaciones
| RREB1 | |
| Polyclonal | |
| Western Blot | |
| DNA-binding protein, FINB, Finger protein in nuclear bodies, hindsight homolog, HNT, LZ321, Raf-responsive zinc finger protein LZ321, ras responsive element binding protein 1, ras-responsive element-binding protein 1, RREB-1, Zep-1, zinc finger motif enhancer binding protein 1, Zinc finger motif enhancer-binding protein 1, zinc finger motif-enhancer binding-protein 1 | |
| The immunogen for this product maps to a region between residue 1375 and 1425 of human Ras Responsive Eement Binding Protein 1 using the numbering given in entry NP_002946.4 (GeneID 6239). | |
| 0.1 mL | |
| Signal Transduction | |
| 6239 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido