missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521392
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, NIH-3T3. Predicted reactivity: Mouse (81%), Dog (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Analysis of cDNA clones indicates that ribosome binding protein 1 may exist in different forms due to removal of tandem repeats, or partial intraexonic splicing of RRBP1. The form presented here is lacking the canine p180 ribosome-binding domain, NQGKKAEGAQ, which is tandemly repeated close to the N-terminus in other forms that haven't been fully characterized. RRBP1 has been excluded as a candidate gene in the cause of Alagille syndrome. Alternate splicing results in multiple transcript variants.
Specifications
| RRBP1 | |
| Polyclonal | |
| Unconjugated | |
| RRBP1 | |
| 1700087N07Rik; 180 kDa ribosome receptor homolog; 5730465C04Rik; ES/130; ES/130-related protein; ES130; hES; KIAA1398; mKIAA1398; mRRp; mRRp0; mRRp1.8; mRRp10; mRRp15a; mRRp15b; mRRp16.8; mRRp2; mRRp41; mRRp47; mRRp5.4; p180; ribosome binding protein 1; ribosome binding protein 1 homolog 180kDa (dog); ribosome receptor protein; Ribosome-binding protein 1; RP11-462D18.3; Rrbp1; RRp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6238, 81910 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
| Q99PL5, Q9P2E9 | |
| RRBP1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Ribosome binding protein 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction