missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-36709-20ul
This item is not returnable.
View return policy
Description
RRBP1 Polyclonal antibody specifically detects RRBP1 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| RRBP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| 180 kDa ribosome receptor homolog, DKFZp586A1420, ES/130, ES/130-related protein, ES130, FLJ36146, hES, KIAA1398, MGC157720, MGC157721, ribosome binding protein 1 (dog 180kD homolog), ribosome binding protein 1 homolog 180kDa (dog), Ribosome receptor protein, ribosome-binding protein 1, RRp | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human RRBP1 (NP_004578.2).,, Sequence:, MDIYDTQTLGVVVFGGFMVVSAIGIFLVSTFSMKETSYEEALANQRKEMAKTHHQKVEKKKKEKTVEKKGKTKKKEEKPNGKIPDHDPAPNVTVLLREPVRAPAVAVAPTPVQPPIIVAPVATVPAMPQEKLASSPKDKK | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6238 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction