missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12530-1-AP-20UL
This item is not returnable.
View return policy
Description
RRAS2 is involved in activating mutations and the overexpression of classical Ras subfamily members (K-RAS, N-RAS and H-RAS) have been widely investigated as key events in the development of human cancers. The RRAS subfamily of Ras-related proteins includes RRAS1, RRAS2 (TC21) and RRAS3 (M-Ras) show overall amino acid identity with the classical Ras subfamily (H-Ras, K-Ras and N-Ras) of 55-60%. RRAS2 is a small GTP binding protein of the Ras superfamily of GTPases. It might transduce growth inhibitory signals across the cell membrane, exerting its effect through an effector shared with the Ras proteins. RRAS2 has high oncogenic potential and overexpression/mutations have been reported in several tumor tissues and cell lines.Specifications
| RRAS2 | |
| Polyclonal | |
| Unconjugated | |
| RRAS2 | |
| Ras like protein TC21, Ras related protein R Ras2, RRAS2, TC21, Teratocarcinoma oncogene | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22800, 66922 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62070, P62071 | |
| Rras2 | |
| RRAS2 Fusion Protein Ag3226 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction