missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRAS2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148732
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
RRAS2 is involved in activating mutations and the overexpression of classical Ras subfamily members (K-RAS, N-RAS and H-RAS) have been widely investigated as key events in the development of human cancers. The RRAS subfamily of Ras-related proteins includes RRAS1, RRAS2 (TC21) and RRAS3 (M-Ras) show overall amino acid identity with the classical Ras subfamily (H-Ras, K-Ras and N-Ras) of 55-60%. RRAS2 is a small GTP binding protein of the Ras superfamily of GTPases. It might transduce growth inhibitory signals across the cell membrane, exerting its effect through an effector shared with the Ras proteins. RRAS2 has high oncogenic potential and overexpression/mutations have been reported in several tumor tissues and cell lines.
Specifications
| RRAS2 | |
| Polyclonal | |
| Unconjugated | |
| RRAS2 | |
| 2610016H24Rik; C86394; Ras-like protein TC21; ras-related protein R-Ras2; related RAS viral (r-ras) oncogene homolog 2; Rras2; TC21; Teratocarcinoma oncogene | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 22800, 365355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P62070 | |
| RRAS2 | |
| E. coli-derived recombinant human TC21/R-Ras2, Met1-Phe204. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction