missing translation for 'onlineSavingsMsg'
Learn More

RRAS Antibody (OTI2B7) [FITC], Novus Biologicals™

Product Code. 30687672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687672 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687672 Supplier Novus Biologicals Supplier No. NBP344965F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RRAS Monoclonal antibody specifically detects RRAS in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAS
Applications Western Blot
Classification Monoclonal
Clone OTI2B7
Conjugate FITC
Formulation PBS
Gene Alias Oncogene RRAS, p23, ras-related protein R-Ras, related RAS viral (r-ras) oncogene homolog
Host Species Mouse
Immunogen Full length human recombiant protein of human RRAS (NP_006261) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6237
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.