missing translation for 'onlineSavingsMsg'
Learn More

RRAS Antibody (2E12), Novus Biologicals™

Product Code. 18392499 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392499 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392499 Supplier Novus Biologicals Supplier No. H00006237M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RRAS Monoclonal antibody specifically detects RRAS in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAS
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 2E12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation 1:10 to 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH16286
Gene Alias Oncogene RRAS, p23, ras-related protein R-Ras, related RAS viral (r-ras) oncogene homolog
Host Species Mouse
Immunogen RRAS (AAH16286, 109 a.a. ~ 218 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AINDRQSFNEVGKLFTQILRVKDRDDFPVVLVGNKADLESQRQVPRSEASAFGASHHVAYFEASAKLRLNVDEAFEQLVRAVRKYQEQELPPSPPSAPRKKGGGCPCVLL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6237
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.