missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRAGD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567003
This item is not returnable.
View return policy
Description
Immunogen sequence: PDSGTEEGVLD FSDPFSTEVK PRILLM Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
RRAGD is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.
Specifications
| RRAGD | |
| Polyclonal | |
| Unconjugated | |
| RRAGD | |
| 5730543C08Rik; AI467523; bA11D8.2.1; C030003H22Rik; D4Ertd174e; rag D; Rag D protein; ragD; Ras related GTP binding D; Ras-related GTP binding D; ras-related GTP-binding protein D; RRAGD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 58528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQL2 | |
| RRAGD | |
| Recombinant Human RRAGD. Recombinant protein control fragment (Product #RP-107644). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction