missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAGD Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32106C
This item is not returnable.
View return policy
Description
RRAGD Polyclonal antibody specifically detects RRAGD in Human, Mouse samples. It is validated for Western Blot
Specifications
| RRAGD | |
| Polyclonal | |
| Western Blot | |
| bA11D8.2.1, DKFZP761H171, FLJ44176, Rag D, Rag D protein, RAGD, Ras-related GTP binding D, ras-related GTP-binding protein D | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Ras-Related GTP Binding D using the numbering given in entry NP_067067.1 (GeneID 58528). | |
| 0.1 mL | |
| Cancer, mTOR Pathway | |
| 58528 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction