missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RRAGD Polyclonal antibody specifically detects RRAGD in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RRAGD |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | bA11D8.2.1, DKFZP761H171, FLJ44176, Rag D, Rag D protein, RAGD, Ras-related GTP binding D, ras-related GTP-binding protein D |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Ras-Related GTP Binding D using the numbering given in entry NP_067067.1 (GeneID 58528). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?